1-Boc-6-hydroxy-1-azaspiro[3.3]heptane 97%
Leverantör: Apollo Scientific
OR317312-1G OR317312-500MG
APOSOR317312-1GEA 17075 SEK
APOSOR317312-1G APOSOR317312-500MG
1-Boc-6-hydroxy-1-azaspiro[3.3]heptane 97%
1-Boc-6-hydroxy-1-azaspiro[3.3]heptane
Formel: C₁₁H₁₉NO₃
MW: 213,273 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD22415220
CAS nummer: 1363381-56-9