4-[(2-Bromo-4,5-dimethoxyphenyl)methyl]-4-[2-[2-(6,6-dimethylbicyclo[3.1.1]hept-2-yl)ethoxy]ethyl]-Morpholinium bromide (1:1) 97%
Leverantör: Apollo Scientific
OR317274-1G OR317274-25G OR317274-5G
APOSOR317274-1GEA 735 SEK
APOSOR317274-1G APOSOR317274-25G APOSOR317274-5G
4-[(2-Bromo-4,5-dimethoxyphenyl)methyl]-4-[2-[2-(6,6-dimethylbicyclo[3.1.1]hept-2-yl)ethoxy]ethyl]-Morpholinium bromide (1:1) 97%
4-[(2-Bromo-4,5-dimethoxyphenyl)methyl]-4-[2-[2-(6,6-dimethylbicyclo[3.1.1]hept-2-yl)ethoxy]ethyl]-Morpholinium bromide (1:1)
Formel: C₂₆H₄₁BrNO₄·Br
MW: 591.416 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01713435
CAS nummer: 53251-94-8