7-Cyano-1,2,3,4-tetrahydroisoquinoline

Leverantör: Apollo Scientific
Varning

OR313006-1G OR313006-250MG
APOSOR313006-1GEA 7325 SEK
APOSOR313006-1G APOSOR313006-250MG
7-Cyano-1,2,3,4-tetrahydroisoquinoline
7-Cyano-1,2,3,4-tetrahydroisoquinoline
Formel: C₁₀H₁₀N₂
Storage Temperature: Omgivande
MDL Number: MFCD03094745
CAS nummer: 149355-52-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR