Bicyclo[1.1.1]pentane-1,3-dicarboxamide
Leverantör: Apollo Scientific
OR312456-1G OR312456-250MG
APOSOR312456-1GEA 1175 SEK
APOSOR312456-1G APOSOR312456-250MG
Bicyclo[1.1.1]pentane-1,3-dicarboxamide
Bicyclo[1.1.1]pentane-1,3-dicarboxamide
Formel: C₇H₁₀N₂O₂
MW: 154.166 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD20716860
CAS nummer: 1379318-53-2