3-Methylbicyclo[1.1.1]pentan-1-amine hydrochloride
Leverantör: Apollo Scientific
OR312452-100MG OR312452-250MG OR312452-1G
APOSOR312452-100MGEA 3225 SEK
APOSOR312452-100MG APOSOR312452-250MG APOSOR312452-1G
3-Methylbicyclo[1.1.1]pentan-1-amine hydrochloride
3-Methylbicyclo[1.1.1]pentan-1-amine hydrochloride
Formel: C₆H₁₁N·ClH
MW: 133.619 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28390442
CAS nummer: 796963-34-3