5-Oxa-2,8-diazaspiro[3.5]nonane, N2-BOC protected
Leverantör: Apollo Scientific
Varning
Synonyms: tert-Butyl 5-oxa-2,8-diazaspiro[3.5]nonane-2-carboxylate
OR312336-1G OR312336-250MG
APOSOR312336-1GEA 11400 SEK
APOSOR312336-1G APOSOR312336-250MG
5-Oxa-2,8-diazaspiro[3.5]nonane, N2-BOC protected
5-Oxa-2,8-diazaspiro[3.5]nonane, N2-BOC protected
Formel: C₁₁H₂₀N₂O₃
MW: 228,288 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD14581202
CAS nummer: 1251011-05-8