N,N,N',N'-Tetrakis(4-aminophenyl)-1,4-phenylenediamine 95%
Leverantör: Apollo Scientific
Synonyms: N1,N1'-(1,4-Phenylene)bis(N1-(4-aminophenyl)benzene-1,4-diamine)
OR3123-1G OR3123-25G OR3123-5G
APOSOR3123-1GEA 1025 SEK
APOSOR3123-1G APOSOR3123-25G APOSOR3123-5G
N,N,N',N'-Tetrakis(4-aminophenyl)-1,4-phenylenediamine 95%
N,N,N',N'-Tetrakis(4-aminophenyl)-1,4-phenylenediamine
Formel: C₃₀H₂₈N₆
MW: 472,584 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06657652
CAS nummer: 3283-07-6