4-Acetyl-N-(2,2-dimethoxyethyl)-3,5-dimethyl-1H-pyrrole-2-carboxamide

Leverantör: Apollo Scientific

OR311028-1G OR311028-500MG
APOSOR311028-1GEA 6350 SEK
APOSOR311028-1G APOSOR311028-500MG
4-Acetyl-N-(2,2-dimethoxyethyl)-3,5-dimethyl-1H-pyrrole-2-carboxamide
4-Acetyl-N-(2,2-dimethoxyethyl)-3,5-dimethyl-1H-pyrrole-2-carboxamide
Formel: C₁₃H₂₀N₂O₄
MW: 268,309 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD14703586
CAS nummer: 1232794-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR