1-(4-Chlorophenyl)-5-methyl-1H-1,2,3-triazole-4-carbohydrazide

Leverantör: Apollo Scientific

OR311026-1G OR311026-500MG
APOSOR311026-1GEA 6350 SEK
APOSOR311026-1G APOSOR311026-500MG
1-(4-Chlorophenyl)-5-methyl-1H-1,2,3-triazole-4-carbohydrazide
1-(4-Chlorophenyl)-5-methyl-1H-1,2,3-triazole-4-carbohydrazide
Formel: C₁₀H₁₀ClN₅O
MW: 251,672 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD14703598
CAS nummer: 242145-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR