3-(2,4-Dimethylphenyl)-2-sulphanylidene-1H,2H,3H,4H-thieno[3,2-d]pyrimidin-4-one

Leverantör: Apollo Scientific

OR311012-1G OR311012-500MG
APOSOR311012-1GEA 6350 SEK
APOSOR311012-1G APOSOR311012-500MG
3-(2,4-Dimethylphenyl)-2-sulphanylidene-1H,2H,3H,4H-thieno[3,2-d]pyrimidin-4-one
3-(2,4-Dimethylphenyl)-2-sulphanylidene-1H,2H,3H,4H-thieno[3,2-d]pyrimidin-4-one
Formel: C₁₄H₁₂N₂OS₂
MW: 288,388 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD17780079
CAS nummer: 811451-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR