4-(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)benzoic acid
Leverantör: Apollo Scientific
Varning
OR310580-5G OR310580-1G
APOSOR310580-5GEA 2800 SEK
APOSOR310580-5G APOSOR310580-1G
4-(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)benzoic acid
4-(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)benzoic acid
Formel: C₁₂H₁₂BNO₆
MW: 277.038 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD26793795
CAS nummer: 1072960-67-8