1,3-Di-tert-butyl 2-methylpropanedioate
Leverantör: Apollo Scientific
OR310525-5G
OR310525-1G
APOSOR310525-5GEA
9325
SEK
APOSOR310525-5G
APOSOR310525-1G
1,3-Di-tert-butyl 2-methylpropanedioate
1,3-Di-tert-butyl 2-methylpropanedioate
Formel:
C₁₂H₂₂O₄ MW: 230,301 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD06797699 CAS nummer: 34812-95-8 |