[(2-Ethylhexyl)oxy]benzene 95%
Leverantör: Apollo Scientific
Varning
OR310479-5G OR310479-1G
APOSOR310479-5GEA 2275 SEK
APOSOR310479-5G APOSOR310479-1G
[(2-Ethylhexyl)oxy]benzene 95%
[(2-Ethylhexyl)oxy]benzene
Formel: C₁₄H₂₂O
MW: 206.324 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD20109150
CAS nummer: 82001-49-8