3-Iodo-5-methyl-1H-pyrazole
Leverantör: Apollo Scientific
|
Varning
|
OR31047-1G
OR31047-5G
APOSOR31047-1GEA
925
SEK
APOSOR31047-1G
APOSOR31047-5G
3-Iodo-5-methyl-1H-pyrazole
3-Iodo-5-methyl-1H-pyrazole
Formel:
C₄H₅IN₂ MW: 208 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD23135760 CAS nummer: 93233-21-7 |