4'-Formyl-2'-nitroacetanilide
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
N-(4-Formyl-2-nitrophenyl)acetamide, 4-Acetamido-3-nitrobenzaldehyde
OR310363-1G
OR310363-5G
APOSOR310363-1GEA
2075
SEK
APOSOR310363-1G
APOSOR310363-5G
4'-Formyl-2'-nitroacetanilide
4'-Formyl-2'-nitroacetanilide
Formel:
C₉H₈N₂O₄ MW: 208,171 g/mol Smältpunkt: 155…156 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00092015 CAS nummer: 51818-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...