1-(6-Chloro-5-methylpyridin-3-yl)ethan-1-one

Leverantör: Apollo Scientific
Varning

Synonyms: 1-(6-Chloro-5-methylpyridin-3-yl)ethanone

OR310313-1G OR310313-100MG OR310313-250MG
APOSOR310313-1GEA 7450 SEK
APOSOR310313-1G APOSOR310313-100MG APOSOR310313-250MG
1-(6-Chloro-5-methylpyridin-3-yl)ethan-1-one
1-(6-Chloro-5-methylpyridin-3-yl)ethan-1-one
Formel: C₈H₈ClNO
MW: 169,608 g/mol
Smältpunkt: 107…109 °C
Storage Temperature: Omgivande
MDL Number: MFCD18258044
CAS nummer: 1256791-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR