5-Chloro-3(3-chlorobenzyl)-1,2,4-thiadiazole
Leverantör: Apollo Scientific
OR309463-100MG
OR309463-250MG
APOSOR309463-100MGEA
1700
SEK
APOSOR309463-100MG
APOSOR309463-250MG
5-Chloro-3(3-chlorobenzyl)-1,2,4-thiadiazole
5-Chloro-3(3-chlorobenzyl)-1,2,4-thiadiazole
Formel:
C₉H₆Cl₂N₂S MW: 245,128 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD14707315 CAS nummer: 1221345-99-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...