2-Chloro-6,7-dimethylquinoline-3-methanol
Leverantör: Apollo Scientific
OR309258-1G OR309258-5G
APOSOR309258-1GEA 3950 SEK
APOSOR309258-1G APOSOR309258-5G
2-Chloro-6,7-dimethylquinoline-3-methanol
2-Chloro-6,7-dimethylquinoline-3-methanol
Formel: C₁₂H₁₂ClNO
Storage Temperature: Omgivande
MDL Number: MFCD02232330
CAS nummer: 333408-44-9