4-Acetyl-1-methyl-1H-indazole
Leverantör: Apollo Scientific
Varning
Synonyms: 4-Ethanoyl-1-methyl-1H-indazole, 1-(1-Methyl-1H-indazol-4-yl)ethanone, 1-(1-Methyl-1H-indazol-4-yl)ethan-1-one
OR30888-1G OR30888-250MG OR30888-500MG
APOSOR30888-1GEA 6575 SEK
APOSOR30888-1G APOSOR30888-250MG APOSOR30888-500MG
4-Acetyl-1-methyl-1H-indazole
4-Acetyl-1-methyl-1H-indazole
Formel: C₁₀H₁₀N₂O
MW: 174,199 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD11869766
CAS nummer: 1159511-23-5