4-(Aminomethyl)-1-methyl-1H-indazole hydrochloride
Leverantör: Apollo Scientific
Varning
Synonyms: (1-Methyl-1H-indazol-4-yl)methanamine hydrochloride, (1-Methyl-1H-indazol-4-yl)methylamine hydrochloride
OR30871-100MG OR30871-250MG OR30871-1G
APOSOR30871-100MGEA 500 SEK
APOSOR30871-100MG APOSOR30871-250MG APOSOR30871-1G
4-(Aminomethyl)-1-methyl-1H-indazole hydrochloride
4-(Aminomethyl)-1-methyl-1H-indazole hydrochloride
Formel: C₉H₁₁N₃·ClH
MW: 197,665 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18803462
CAS nummer: 1334405-46-7