2-Chloro-6,8-dimethylquinoline-3-carboxaldehyde
Leverantör: Apollo Scientific
OR307916-1G OR307916-5G
APOSOR307916-1GEA 3050 SEK
APOSOR307916-1G APOSOR307916-5G
2-Chloro-6,8-dimethylquinoline-3-carboxaldehyde
2-Chloro-6,8-dimethylquinoline-3-carboxaldehyde
Formel: C₁₂H₁₀ClNO
MW: 219,667 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02227044
CAS nummer: 73568-31-7