Methyl diexo-3-Amino-7-oxa-bicyclo[2.2.1]heptane-2-carboxylate hydrochloride
Leverantör: Apollo Scientific
OR307668-1G OR307668-250MG
APOSOR307668-1GEA 7850 SEK
APOSOR307668-1G APOSOR307668-250MG
Methyl diexo-3-Amino-7-oxa-bicyclo[2.2.1]heptane-2-carboxylate hydrochloride
Methyl diexo-3-Amino-7-oxa-bicyclo[2.2.1]heptane-2-carboxylate hydrochloride
Formel: C₈H₁₃NO₃·ClH
MW: 207,655 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09997948
CAS nummer: 4505-16-2