3-(9-H-Fluoren-9-ylmethoxycarbonylamino)-heptanoic acid
Leverantör: Apollo Scientific
OR307655-1G
OR307655-250MG
APOSOR307655-1GEA
8725
SEK
APOSOR307655-1G
APOSOR307655-250MG
3-(9-H-Fluoren-9-ylmethoxycarbonylamino)-heptanoic acid
3-(9-H-Fluoren-9-ylmethoxycarbonylamino)-heptanoic acid
Formel:
C₂₂H₂₅NO₄ MW: 367,438 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD03412497 CAS nummer: 683219-85-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...