cis-2-Amino-cyclohex-3-enecarboxylic acid hydrochloride
Leverantör: Apollo Scientific
OR307645-1G OR307645-250MG
APOSOR307645-1GEA 7150 SEK
APOSOR307645-1G APOSOR307645-250MG
cis-2-Amino-cyclohex-3-enecarboxylic acid hydrochloride
cis-2-Amino-cyclohex-3-enecarboxylic acid hydrochloride
Formel: C₇H₁₁NO₂·ClH
MW: 177.629 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08669705
CAS nummer: 142035-00-5