3-(Bromomethyl)-1H-indole, N-BOC protected 97%
Leverantör: Apollo Scientific
Varning
Synonyms: tert-Butyl 3-(bromomethyl)-1H-indole-1-carboxylate, 3-(Bromomethyl)-1-(tert-butoxycarbonyl)-1H-indole
OR30757-1G OR30757-5G
APOSOR30757-1GEA 1125 SEK
APOSOR30757-1G APOSOR30757-5G
3-(Bromomethyl)-1H-indole, N-BOC protected 97%
3-(Bromomethyl)-1H-indole, N-BOC protected
Formel: C₁₄H₁₆BrNO₂
Smältpunkt: 106 °C
Storage Temperature: Kylskåp
MDL Number: MFCD05864361
CAS nummer: 96551-21-2