4-Chloro-1-(1-ethoxyethyl)-3-iodo-1H-pyrazole
Leverantör: Apollo Scientific
Varning
OR307300-1G OR307300-250MG
APOSOR307300-1GEA 4275 SEK
APOSOR307300-1G APOSOR307300-250MG
4-Chloro-1-(1-ethoxyethyl)-3-iodo-1H-pyrazole
4-Chloro-1-(1-ethoxyethyl)-3-iodo-1H-pyrazole
Formel: C₇H₁₀ClIN₂O
MW: 300,525 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD31537585
CAS nummer: 2197054-81-0