3-Bromo-4-nitro-1H-pyrazole
Leverantör: Apollo Scientific
Varning
OR307162-1G OR307162-250MG OR307162-5G
APOSOR307162-1GEA 2450 SEK
APOSOR307162-1G APOSOR307162-250MG APOSOR307162-5G
3-Bromo-4-nitro-1H-pyrazole
3-Bromo-4-nitro-1H-pyrazole
Formel: C₃H₂BrN₃O₂
MW: 191,971 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD19441764
CAS nummer: 784193-37-9