2H-Spiro[1-benzofuran-3,4'-piperidine] 97%
Leverantör: Apollo Scientific
Synonyms: 2H-Spiro[benzofuran-3,4'-piperidine]
OR306350-1G OR306350-250MG
APOSOR306350-1GEA 13675 SEK
APOSOR306350-1G APOSOR306350-250MG
2H-Spiro[1-benzofuran-3,4'-piperidine] 97%
2H-Spiro[1-benzofuran-3,4'-piperidine]
Formel: C₁₂H₁₅NO
MW: 189.254 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD20488777
CAS nummer: 171-77-7