Methyl (5-butyl-1H-benzimidazol-2-yl)carbamate 95%
Leverantör: Apollo Scientific
Varning
Synonyms: Methyl (5-butyl-1H-benzo[d]imidazol-2-yl)carbamate, Parbendazole
OR305450-1G OR305450-100MG OR305450-250MG
APOSOR305450-1GEA 2375 SEK
APOSOR305450-1G APOSOR305450-100MG APOSOR305450-250MG
Methyl (5-butyl-1H-benzimidazol-2-yl)carbamate 95%
Methyl (5-butyl-1H-benzimidazol-2-yl)carbamate
Formel: C₁₃H₁₇N₃O₂
MW: 247,293 g/mol
Smältpunkt: 255…257 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00864534
CAS nummer: 14255-87-9
EINECS: 238-133-3