5-Nitroisoindolin-1-one
Leverantör: Apollo Scientific
Varning
Synonyms: 2,3-Dihydro-5-nitro-1H-isoindol-1-one, 2,3-Dihydro-5-nitro-1-oxo-1H-isoindole
OR305280-1G OR305280-250MG OR305280-5G
APOSOR305280-1GEA 2025 SEK
APOSOR305280-1G APOSOR305280-250MG APOSOR305280-5G
5-Nitroisoindolin-1-one
5-Nitroisoindolin-1-one
Formel: C₈H₆N₂O₃
MW: 178.145 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD10000827
CAS nummer: 876343-38-3