4-Methoxybenzene-1,2-diol 95%
Leverantör: Apollo Scientific
Varning
Synonyms: 3,4-Dihydroxyanisole, 4-Methoxycatechol
OR305004-1G OR305004-250MG OR305004-100MG
APOSOR305004-1GEA 4550 SEK
APOSOR305004-1G APOSOR305004-250MG APOSOR305004-100MG
4-Methoxybenzene-1,2-diol 95%
4-Methoxybenzene-1,2-diol
Formel: C₇H₈O₃
MW: 140,137 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18395726
CAS nummer: 3934-97-2