2-(3,3-Dimethoxycyclobutyl)acetonitrile

Leverantör: Apollo Scientific

OR304371-1G OR304371-250MG OR304371-500MG
APOSOR304371-1GEA 9300 SEK
APOSOR304371-1G APOSOR304371-250MG APOSOR304371-500MG
2-(3,3-Dimethoxycyclobutyl)acetonitrile
2-(3,3-Dimethoxycyclobutyl)acetonitrile
Formel: C₈H₁₃NO₂
MW: 155.194 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD30003025
CAS nummer: 2007916-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR