3-(Propan-2-yl)azetidin-3-ol hydrochloride
Leverantör: Apollo Scientific
OR304332-1G OR304332-5G
APOSOR304332-1GEA 9550 SEK
APOSOR304332-1G APOSOR304332-5G
3-(Propan-2-yl)azetidin-3-ol hydrochloride
3-(Propan-2-yl)azetidin-3-ol hydrochloride
Formel: C₆H₁₃NO·ClH
MW: 151.634 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD27959490
CAS nummer: 848192-94-9