(2S)-1,3-Dimethylazetidine-2-carboxylic acid
Leverantör: Apollo Scientific
OR304328-1G
OR304328-250MG
OR304328-500MG
APOSOR304328-1GEA
20900
SEK
APOSOR304328-1G
APOSOR304328-250MG
APOSOR304328-500MG
(2S)-1,3-Dimethylazetidine-2-carboxylic acid
(2S)-1,3-Dimethylazetidine-2-carboxylic acid
Formel:
C₆H₁₁NO₂ MW: 129,157 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD30146476 CAS nummer: 1860067-54-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...