(2S)-1-Ethylazetidine-2-carboxylic acid

Leverantör: Apollo Scientific

OR304326-1G OR304326-5G
APOSOR304326-1GEA 7725 SEK
APOSOR304326-1G APOSOR304326-5G
(2S)-1-Ethylazetidine-2-carboxylic acid
(2S)-1-Ethylazetidine-2-carboxylic acid
Formel: C₆H₁₁NO₂
MW: 129,157 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18835081
CAS nummer: 255882-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR