1-(Azetidin-3-yl)ethan-1-ol hydrochloride
Leverantör: Apollo Scientific
OR304324-1G OR304324-250MG OR304324-500MG
APOSOR304324-1GEA 12175 SEK
APOSOR304324-1G APOSOR304324-250MG APOSOR304324-500MG
1-(Azetidin-3-yl)ethan-1-ol hydrochloride
1-(Azetidin-3-yl)ethan-1-ol hydrochloride
Formel: C₅H₁₁NO·ClH
MW: 137.608 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD29918536
CAS nummer: 2068152-34-9