Methyl 4'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-3-carboxylate
Leverantör: Apollo Scientific
OR303563-1G OR303563-500MG OR303563-5G
APOSOR303563-1GEA 1825 SEK
APOSOR303563-1G APOSOR303563-500MG APOSOR303563-5G
Methyl 4'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-3-carboxylate
Methyl 4'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-3-carboxylate
Formel: C₂₀H₂₃BO₄
Storage Temperature: Omgivande
MDL Number: MFCD16294535
CAS nummer: 1256358-85-6