N-Acetylcytidine
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
N-Acetyl-1-(β-D-ribofuranosyl)cytosine, N-{1,2-Dihydro-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl]-2-oxopyrimidin-4-yl}acetamide, N-Ethanoylcytidine, N-[1,2-Dihydro-2-oxo-1-(β-D-ribofuranosyl)pyrimidin-4-yl]acetamide, N4-Acetylcytidine
OR303359-10G
OR303359-1G
OR303359-5G
APOSOR303359-10GEA
1050
SEK
APOSOR303359-10G
APOSOR303359-1G
APOSOR303359-5G
N-Acetylcytidine
N-Acetylcytidine
Formel:
C₁₁H₁₅N₃O₆ MW: 285,253 g/mol Smältpunkt: 199 °C (dec.) Storage Temperature: Frys |
MDL Number:
MFCD00006540 CAS nummer: 3768-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...