Ethyl 4-ethyl-1,3-thiazole-2-carboxylate

Leverantör: Apollo Scientific

OR303244-1G OR303244-500MG
APOSOR303244-1GEA 3850 SEK
APOSOR303244-1G APOSOR303244-500MG
Ethyl 4-ethyl-1,3-thiazole-2-carboxylate
Ethyl 4-ethyl-1,3-thiazole-2-carboxylate
Formel: C₈H₁₁NO₂S
MW: 185,243 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11520453
CAS nummer: 79247-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR