4,6-Dichloro-N-methoxy-N-methylpyrimidine-5-carboxamide
Leverantör: Apollo Scientific
OR303235-1G OR303235-500MG
APOSOR303235-1GEA 7900 SEK
APOSOR303235-1G APOSOR303235-500MG
4,6-Dichloro-N-methoxy-N-methylpyrimidine-5-carboxamide
4,6-Dichloro-N-methoxy-N-methylpyrimidine-5-carboxamide
Formel: C₇H₇Cl₂N₃O₂
Storage Temperature: Omgivande
MDL Number: MFCD23697509
CAS nummer: 1426904-70-2