2-(4-Bromophenyl)-4,5,6,7-tetrahydrofuro[3,2-c]pyridine hydrochloride
Leverantör: Apollo Scientific
OR303153-1G OR303153-500MG
APOSOR303153-1GEA 2575 SEK
APOSOR303153-1G APOSOR303153-500MG
2-(4-Bromophenyl)-4,5,6,7-tetrahydrofuro[3,2-c]pyridine hydrochloride
2-(4-Bromophenyl)-4,5,6,7-tetrahydrofuro[3,2-c]pyridine hydrochloride
Formel: C₁₃H₁₂BrNO·ClH
MW: 314,605 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD22689165
CAS nummer: 104278-48-0