1-(Azetidin-3-yl)pyrrolidin-2-one hydrochloride
Leverantör: Apollo Scientific
OR302320-1G OR302320-250MG
APOSOR302320-1GEA 13650 SEK
APOSOR302320-1G APOSOR302320-250MG
1-(Azetidin-3-yl)pyrrolidin-2-one hydrochloride
1-(Azetidin-3-yl)pyrrolidin-2-one hydrochloride
Formel: C₇H₁₂N₂O·ClH
MW: 176.644 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD22581550
CAS nummer: 1403766-91-5