5-[3-(tert-Butyl)-1-(2,4-dichlorobenzyl)-1H-pyrazol-5-yl]-4-methyl-4H-1,2,4-triazole-3-thiol
Leverantör: Apollo Scientific
OR30053-10G OR30053-1G OR30053-5G
APOSOR30053-10GEA 5100 SEK
APOSOR30053-10G APOSOR30053-1G APOSOR30053-5G
5-[3-(tert-Butyl)-1-(2,4-dichlorobenzyl)-1H-pyrazol-5-yl]-4-methyl-4H-1,2,4-triazole-3-thiol
5-[3-(tert-Butyl)-1-(2,4-dichlorobenzyl)-1H-pyrazol-5-yl]-4-methyl-4H-1,2,4-triazole-3-thiol
Formel: C₁₇H₁₉Cl₂N₅S
MW: 396.337 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01765440
CAS nummer: 306937-18-8