1-Chloroisoquinoline-7-carboxaldehyde
Leverantör: Apollo Scientific
Varning
OR300075-250MG OR300075-100MG
APOSOR300075-250MGEA 2550 SEK
APOSOR300075-250MG APOSOR300075-100MG
1-Chloroisoquinoline-7-carboxaldehyde
1-Chloroisoquinoline-7-carboxaldehyde
Formel: C₁₀H₆ClNO
Storage Temperature: Kylskåp
MDL Number: MFCD09835329
CAS nummer: 223671-53-2