Naphthalene-2-sulphonic acid hydrate

Leverantör: Apollo Scientific

Synonyms: 2-Naphthalenesulphonic acid hydrate

OR28837-1KG OR28837-250G OR28837-50G
APOSOR28837-1KGEA 5975 SEK
APOSOR28837-1KG APOSOR28837-250G APOSOR28837-50G
Naphthalene-2-sulphonic acid hydrate
Naphthalene-2-sulphonic acid hydrate
Formel: C₁₀H₈O₃S·xH₂O
MW: 208,231 g/mol (anhy)
Smältpunkt: 124 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149271
CAS nummer: 6036-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR