3-Hydroxybutanenitrile

Leverantör: Apollo Scientific
Varning

Synonyms: 1-Cyanopropan-2-ol, 3-Hydroxybutyronitrile

OR2847-1G OR2847-250MG
APOSOR2847-1GEA 3775 SEK
APOSOR2847-1G APOSOR2847-250MG
3-Hydroxybutanenitrile
3-Hydroxybutanenitrile
Formel: C₄H₇NO
MW: 85,104 g/mol
Kokpunkt: 214 °C
Densitet: 0,976 g/cm³ (25 °C)
Flampunkt: 110 °C
Storage Temperature: Omgivande
MDL Number: MFCD03427120
CAS nummer: 4368-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR