Methyl 2,4-dioxo-4-(2-nitrophenyl)butanoate
Leverantör: Apollo Scientific
Varning
OR28131-100MG OR28131-250MG OR28131-500MG
APOSOR28131-100MGEA 2475 SEK
APOSOR28131-100MG APOSOR28131-250MG APOSOR28131-500MG
Methyl 2,4-dioxo-4-(2-nitrophenyl)butanoate
Methyl 2,4-dioxo-4-(2-nitrophenyl)butanoate
Formel: C₁₁H₉NO₆
Storage Temperature: Omgivande
MDL Number: MFCD00832016
CAS nummer: 57446-03-4