Diethyl 2-heptylmalonate
Leverantör: Apollo Scientific
Varning
OR27453-1G OR27453-100MG OR27453-250MG OR27453-500MG OR27453-2.5G OR27453-5G
APOSOR27453-1GEA 4800 SEK
APOSOR27453-1G APOSOR27453-100MG APOSOR27453-250MG APOSOR27453-500MG APOSOR27453-2.5G APOSOR27453-5G
Diethyl 2-heptylmalonate
Diethyl 2-heptylmalonate
Formel: C₁₄H₂₆O₄
Storage Temperature: Omgivande
MDL Number: MFCD00027827
CAS nummer: 607-83-0