1-Chloro-6,6-dimethyl-3-(methylthio)-4,5,6,7-tetrahydrobenzo[c]thiophen-4-one
Leverantör: Apollo Scientific
OR25412-10G OR25412-1G OR25412-5G
APOSOR25412-10GEA 5600 SEK
APOSOR25412-10G APOSOR25412-1G APOSOR25412-5G
1-Chloro-6,6-dimethyl-3-(methylthio)-4,5,6,7-tetrahydrobenzo[c]thiophen-4-one
1-Chloro-6,6-dimethyl-3-(methylthio)-4,5,6,7-tetrahydrobenzo[c]thiophen-4-one
Formel: C₁₁H₁₃ClOS₂
MW: 260,803 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00108830
CAS nummer: 175202-90-1