Ethyl 3-(isopropylthio)-6,6-dimethyl-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylate
Leverantör: Apollo Scientific
OR25272-1G OR25272-5G
APOSOR25272-1GEA 855 SEK
APOSOR25272-1G APOSOR25272-5G
Ethyl 3-(isopropylthio)-6,6-dimethyl-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylate
Ethyl 3-(isopropylthio)-6,6-dimethyl-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylate
Formel: C₁₆H₂₂O₃S₂
Storage Temperature: Omgivande
MDL Number: MFCD00085069
CAS nummer: 175202-44-5